Learn More
TARGETMOL CHEMICALS INC PHA 543613 5MG
Supplier: TARGETMOL CHEMICALS INC T23145
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. PHA 543613 is a specific agonist of α7 nAChR (Ki = 8.8 nM) and can be used in studies about the cognitive deficits of Alzheimer's disease and schizophrenia. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.