Learn More
Chemglass Life Sciences pH Probe, with VP, 12 x 425mm
Supplier: Chemglass Life Sciences CLS143537
pH probe with VP, 12x425mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.