Learn More
Chemglass Life Sciences pH Probe, with VP, 12 x 120mm
SDP
Supplier:  Chemglass Life Sciences CLS143530
Encompass_Preferred
pH probe with VP, 12x120mm. • pH Range: 0-12 • Electrodes are filled with gel reference electrolyte under a pressure of 2.5 bar • Pressure Range: From vacuum up to a process of maximum 36PSI (2.5 bar) • Temperature Range: Steam sterilization/autoclavable to 266°F (0 to 130°C) • In-situ sterilization: Up to 100 cycles under standard conditions • Fully autoclavable or in-situ sterializable
Catalog No. 50-143-149
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.