Learn More
Chemglass Life Sciences PH Electrodes for Chemglass Reactors, 12mm Shaft Diameter, 450mm Usable Shaft Length, 20mm Minimum Immersion Length, Glass Shaft Material, Fixed Cable Electrode Head, 0 to 100°C Temperature Range
SDP
Supplier:  Chemglass Life Sciences CG187202
Encompass_Preferred
Metrohm pH electrodes designed to be used with Chemglass reactors and data loggers. All electrodes are supplied with integrated PT 1000 sensor for temperature correction and have a BNC connector. 12mm Shaft Diameter, 450mm Usable Shaft Length, 20mm Minimum Immersion Length, Glass Shaft Material, Fixed Cable Electrode Head, 0 to 100°C Temperature Range.12mm OD probes designed to go directly into smaller reactors using the standard taper joint adapters (CG-1872-A-01 through CG-1872-A-03).
Catalog No. CG187202
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.