Learn More
Eurofins Discoverx PathHunter PL/PK Detection kit, 4,000 dp (384-well). (93-0812)
Supplier: Eurofins Discoverx 930812
The PathHunter ProLabel/PK Detection kit is a detection kit intended for use with Eurofins DiscoverX's PathHunter cell lines or with custom cell lines overexpressing ED (ProLabel/PK) but not containing any EA. This kit contains lysis buffer, exogenous EA, and EFC substrate to allow for detection of levels of ED in the assayed cells. This kit contains all components required to assay 10 plates (96/384-well). For detailed product information, User manual and COA, visit www.discoverx.com/products/reagents/detection-kit-93-0812.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.