Learn More
Hach Company pH Paper, 6.0 - 8.0 pH Range
Supplier:  Hach Company 37933
Encompass_Preferred
Provides simple, fast, economical method of pH measurement. Sharp color change for each 0.5 pH unit. Can be used with handy dispenser (Product No. 2611000). Convenient for sample pH adjustments. Recommended for sewage water, not finished water. Five 4.6-meter (15') rolls per package.
Catalog No. 50-199-4265
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.