Learn More
CTL Scientific Supply Corp PH-FIX STR. 6-7.7,CS/10BX/100
SDP
Supplier:  CTL Scientific Supply Corp 9215010
Encompass_Preferred
PH-FIX 6.0-7.7 TEST PAPER are non-bleeding indicator strips. Gradation of scale: 6.0 - 6.4 - 6.7 - 7.0 - 7.3 - 7.7 (case of 10 boxes of 100 strips each)
Catalog No. 50-117-3350
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.