Learn More
New England Biolabs, Inc. Ph.D.™-12 Phage Display Peptide Library – 50 panning experiments
Supplier: New England Biolabs, Inc. E8111L
The Ph.D.-12 Phage Display Peptide Library contains a single tube of the Ph.D. -12 Phage Library. The Ph.D.-12 library is a combinatorial library of random 12-mer peptides fused to a minor coat protein (pIII) of M13 phage. The displayed peptide is expressed at the N-terminus of pIII. The library consists of approximately 1,000,000,000 electroporated (i.e., unique) sequences.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.