Learn More
Chemglass Life Sciences Complete Glass Baffles (For 10L-20L) Reactors: Overall Baffle Length: 32"", Effective Baffle Length: 13.5"", Baffle Wxd: 19X19Mm, Lower Compression Tfe Fitting: 1/2"", Adapter Joint Size: 45/50
Supplier: Chemglass Life Sciences CG1872B02
Complete Glass Baffles for use in 10L-20L Reactors: Overall Baffle Length: 32"""", Effective Baffle Length: 13.5"""", Baffle Width x Depth (mm): 19 x 19, Lower Compression TFE Fitting: 1/2"""", Adapter Joint Size: 45/50.Features: • Available in Square or Rectangular Shapes, with Open Bottom • PTFE Joint is Offset Permitting Baffle to be Adjusted up to 1"""" Towards the Center of the Reactor • Baffle Can Be Vertically Adjusted Inside Reactor • Hollow Baffles Allow pH Electrodes to be Inserted Into the Reaction Complete Baffles Consist of: Glass Baffle, Glass Fitted PTFE Adapter, and Lower PTFE Compression Fitting.Unique glass baffle system for process reactors from 2L to 50L. Designed to increase agitation in the reactor and mimic baffles found in larger glass lined reactors. All baffles have 3/4"""" precision ground top and lower tubing for use with compression fittings and top jointed adapter.†Designed and developed by John Korzun, Bristol Myers Squibb, New Brunswick, NJ
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.