Learn More
Hach Company Titration Application Pack Water: pH/EC & Alkalinity
Supplier:  Hach Company AP0002.AT1102
Encompass_Preferred
Application Pack for the determination of pH/EC and Alkalinity in water. Suited for TitraLab AT1000 Series, Automatic Titrator Models AT1102/AT1112/AT1122/AT1222.
Catalog No. 50-199-7507
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.