Learn More
Enzo Life Sciences PGF2alpha high sensitivity ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI931069
Encompass_Preferred
The PGF2α high sensitivity EIA kit is a colorimetric competitive enzyme immunoassay kit with results overnight + 3 hours. Absorbance is read at 405 nm. Simply the most sensitive quantification fo PGF2α commercially available in an enzyme immunoassay. Assay more samples without extraction. Alternative name: Prostaglandin F2alpha. Assay Time: Overnight + 3 hours. Applications: ELISA, Colorimetric detection . Species reactivity: Species independent .
Catalog No. 50-200-7110
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.