Learn More
Enzo Life Sciences PGE2 ELISA kit (5x96wells)
SDP
Supplier:  Enzo Life Sciences ADI901001
Encompass_Preferred
Highly sensitive PGE2 ELISA kit for inflammation and eicosanoid research. The PGE2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 2. Commercially available since 1992, this kit is widely cited in peer-reviewed publications. The non-radioactive ready-to-use liquid color-coded reagents reduce errors. Alternative Name: Prostaglandin E2. Sensitivity: 13.4 pg/ml (range 39.1 - 2,500 pg/ml). Assay Time: <3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-197
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.