Learn More
Andwin Scientific Petri Dish Labels, 50 square grid label for petri dishes, 36/pk
SDP
Supplier:  Andwin Scientific PSTK1050
Encompass_Preferred
Made with water resistant ink substrate and adhesive. Will withstand temperatures from 42 deg C to 4 deg C. Apply to back of petri dish. 3 inch (76mm) diameter.
Catalog No. NC1663650
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.