Learn More
Bellco Glass, Inc. Ideal for spreading Inoculum over agar face in Petri dish cultures. Manufactured from Type 1 Class A Borosilicate Glass Rod. Length: 130mm Style bend: 120 degrees
Supplier: Bellco Glass, Inc. 565400120
Ideal for spreading Inoculum over agar face in Petri dish cultures. Manufactured from Type 1 Class A Borosilicate Glass Rod. Length: 130mm Style bend: 120 degrees