Learn More
Enzo Life Sciences Penitrem A (5mg). CAS: 12627-35-9
SDP
Supplier:  Enzo Life Sciences BMLKC1570005
Encompass_Preferred
Fungal mycotoxin. Selective, irreversible blocker of the smooth muscle high conductance Ca2+-activated K+ (maxi-K) channel (100% block at 10 nM). Increases spontaneous release of GABA and aspartate from cerebrocortical synaptosomes in rat neuromuscular junction. Alternative name: NSC 354845, Tremortin A. Purity: ≥95% (HPLC, TLC). Solubility: Soluble in organic solvents such as methanol, acetone or DMSO. Long Term Storage: -20°C.
Catalog No. 50-201-1070
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.