Learn More
CTL Scientific Supply Corp PEHANON INDICATOR STRIPS 2.8-4
Supplier: CTL Scientific Supply Corp 90414
Pehannon Indicator Strips where the indicator and color scale are combined on a single strip. Gradation of scale: 2.8 - 3.1 - 3.4 - 3.6 - 3.8 - 4.0 - 4.3 - 4.6 200 strips/box