Learn More
CTL Scientific Supply Corp PEHANON INDICATOR STRIPS 1.0-
Supplier: CTL Scientific Supply Corp 90412
Pehannon Indicator Strips where the indicator and color scale are combined on a single strip. Gradation of scale: 1.0 - 1.3 -1.6 - 1.8 - 2.0 - 2.2 - 2.5 - 2.8 200 strips/box