Learn More
Electron Microscopy Sciences Peel Away Disposable Embedding Mold Assorted Sample Pack
Supplier: Electron Microscopy Sciences 70180AS
Peel Away Disposable Embedding Molds are used for plastic or paraffin embedding. No block trimming needed. Inside fingers hold ID slip which becomes embedded in the block for positive permanent identification.