Learn More
Biolegend PE/Cyanine7 anti-human CD235a
Supplier: Biolegend 349111
PE/Cyanine7 anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests