Learn More
Andwin Scientific PCR 8-Well Tube Strips, 0.2 mL, Natural, 125/pk
Supplier: Andwin Scientific 82006606
Tube strips with attached bubble cap strips. Ideal for all PCR protocols, these 0.1 and 0.2 mL tubes and caps are made from polypropylene and are compatible with most popular thermal cycler blocks. Tube strips are certified free of RNase, DNase, and human DNA (lot-by-lot certificates available).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.