Learn More
Andwin Scientific PATHCO BLADE HANDLE
Supplier: Andwin Scientific D2877
The razor blade is specially made to efficiently aid in cutting tissues. The blade is disposable and is measured 2 inches long. It is made of hard anodized aluminum and is autoclavable. A handle is included with this product.