Learn More
Electron Microscopy Sciences Paraplast Regular 1 KG
Supplier:  Electron Microscopy Sciences 19218
Encompass_Preferred
Handy pellet Paraplast melts rapidly. Melting Point 56 degree C. Cuts to 4 micron thickness with excellent ribbon continuity - no Crumbling or cracking. Double filtered. No need for filtration before use.
Catalog No. 50-276-91
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.