Learn More
Electron Microscopy Sciences Paramat 10 KG
Supplier: Electron Microscopy Sciences 1928610
Paramat is the original British blend of paraffin wax and plastic polymers and it has been used in histology labs throughout the world for over 20 years. It has been found that when plastic polymers are added to the paraffin the elasticity of the final block is greater as compared with paraffin alone. As well the mixture offers improved tissue penetration, easier ribboning of sections, reduced tendency to crumble, and improved overall results with fibrous tissue.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.