Learn More
Electron Microscopy Sciences Paraformaldehyde 4% in 0.2M Phosphate Buffer, pH 7.4 10 ML
Supplier:  Electron Microscopy Sciences 1573595
Encompass_Preferred
Paraformaldehyde fixatives are prepared fresh upon receipt of order to ensure maximum shelf life. In 0.2M Phosphate Buffer. Filled in 20 ml glass vials (10 ml/vial) allowing for fixation of your specimens without transfer.
pH: 7.4
Catalog No. 50-365-974
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.