Learn More
Epredia™ Paraffin Wax Dispenser
Ultra-fast heating system minimizes the wait time for molten wax. Non-drip, three position tap includes a dedicated tap-heating system that ensures even-flow of paraffin wax with no blockages.
Supplier:  Epredia™ A84600011
 View more versions of this product
Catalog No. A84600011
Description
Description
  • Digital temperature control
  • Aluminum inner tank for temperature control
  • Insulated outer tank prevents heat loss
  • Anti-microbial coating
Specifications
Specifications
14 kg
42.1 cm
36.2 cm
50/60 Hz
15 L
43.6 cm
110/120 V
High-Capacity Wax Dispenser
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.