Learn More
Enzo Life Sciences PAPA NONOate (5mg). CAS: 146672-58-4
SDP
Supplier:  Enzo Life Sciences ALX4300165005
Encompass_Preferred
Nitric oxide (NO) donor. The half-life time of PAPA NONOate is 76.6 min. at 22°C in 0.1M phosphate buffer, pH 7.4. Decomposition of NONOates is nearly instantaneous at pH 5.0. Alternative name: PAPA/NO, NOC-15, (Z)-1-[N-(3-Ammoniopropyl)-N-(n-propyl)amino]diazen-1-ium-1,2-diolate. Purity: ≥97% (TLC). Solubility: Soluble in water or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9144
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.