Learn More
Andwin Scientific PALL 25MM ACRODISC .45UMFILTR
Supplier: Andwin Scientific 28143292 CS
The 25 mm Acrodisc� Ion Chromatography Syringe Filters are ideal tools for chromatography. These filters offer consistency of produced results and reliability with its capacity of carrying a liquid volume of up to 125�L. It is sold in a case of 200.