Learn More
Wilmad Labglass PACKING SOLID GLASS BEADS 6MM
Supplier: Wilmad Labglass LG6750106
Distillation Column Packing, Solid Glass Beads. Approximate Diameter: 6mm; Approximate Number Per Pound: 1,600. Manufactured from borosilicate glass. Sold in one pound packages.