Learn More
Eurofins Discoverx PACAP (1-27), 0.5 mg. (92-1061)
Supplier:  Eurofins Discoverx 921061
Encompass_Preferred
Pituitary adenylate cyclase-activating polypeptide 27 (PACAP-27) is an endogenous neuropeptide that stimulates adenylate cyclase and increases cAMP levels. It acts as a neurotransmitter and neuromodulator, and demonstrates stimulatory effects on sympathetic neuronal NPY and catecholamine production. Has considerable homology with vasoactive intestinal peptide (VIP). Acts as an agonist for ADCYAP1R1. Provided as 500 μg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9602
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.