Learn More
Enzo Life Sciences PAC-1 (10mg). CAS: 315183-21-2
SDP
Supplier:  Enzo Life Sciences ALX430146M010
Encompass_Preferred
First molecule to directly activate procaspase-3 to caspase-3 in vitro. Has been shown to induce apoptosis in cancer cells. Alternative name: 4-(Phenylmethyl)[[2-hydroxy-3-(2-propenyl)phenyl]methylene]hydrazide-1-piperazineacetic acid. Purity: ≥98% (HPLC (UV)). Solubility: Soluble in acetonitrile, DMSO, dioxane or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-9234
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.