Learn More
Enzo Life Sciences p62 ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI9002120001
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Highly validated, quantitative p62 ELISA kit available for autophagy research. The p62 ELISA kit is a colorimetric, immunometric immunoassay kit with results in 3 hours. Alternative Name: Sequestosome 1. Sensitivity: 100pg/ml  (range 625 - 40,000pg/ml). Assay Time: 3 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Human, Mouse, Rat. Contents: Microtiter Plate, Antibody, Conjugate, Assay Buffer 13, Wash Buffer Concentrate, Standard, TMB Substrate, Stop Solution 2, RIPA Cell Lysis Buffer 2
Catalog No. NC0291321