Learn More
Lonza Walkersville P3 PRIMARY CELL KIT 2 X 384RXN
Supplier:  Lonza Walkersville V5SP3002
Encompass
P3 Primary Cell 384-well Nucleofector™ KitFor high-throughput transfection of primary cells, e.g. human T cells, DCs or CD34, when working with the 384-well HT NucleofectorTM System.The P3 Primary Cell 384-well NucleofectorTM Kit is one of our five kits suited for high-throughput transfection of primary cells when working with the 384-well HT NucleofectorTM System.This kit has been determined to be optimal for transfecting following cell types:B cell (human)CD34+ cell (human)Chondrocyte (human)DC (human)DC (mouse), matureES (human), H9ES (mouse)Hepatocyte (human)HMEC (human)Keratinocyte (human)Macrophage (human)Monocyte (human)Neuron (rat), brain Hi/CxNHBE (human)RPE (human)T cell (human), stim.T cell (mouse), unstim.
Catalog No. NC2306205
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.