Learn More
Lonza Walkersville P3 Primary Cell 4D-Nucleofector X Kit S
Supplier: Lonza Walkersville V4XP3032
For efficient transfection of specific primary cells, e.g. human T cells, CD34 or rat neurons, in the 4D-NucleofectorTM X Unit (20 µL format). -High transfection efficiencies for primary cells. -Cells maintain functionality post transfection. -Conditions are transferable between 4D-NucleofectorTM System, 96-well ShuttleTM System and 384-well NucleofectorTM System. Content & Storage: 2 x 0.675 mL P3 Primary Cell NucleofectorTM Solution.2 x 0.15 mL Supplement 1.50 µg pmaxGFPTM Vector (1 μg/μl in 10 mM Tris pH 8.0).2 x 16-well NucleocuvetteTM Strips (20 μl).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.