Learn More
Lonza Walkersville P3 Primary Cell 4D-Nucleofector LV Kit XL
Supplier: Lonza Walkersville V4LP3020
The P3 Primary Cell 4D-Nucleofector LV Kit XL* is suited for larger scale transfection of primary cells in the LV Nucleocuvette Cartridge format when working with the 4D-Nucleofector LV Unit. With the LV Nucleocuvette Cartridge a variable volume of up to 20 mL can be transfected. The cartridge is automatically filled and emptied with help of the supplied input and output reservoirs. Alternatively bags can be used. An optional second input allows for separate feeding of mRNA to avoid its degradation.Applications:For large scale transfection of up to 1 x 10⁹ cellsEx-vivo modification of human primary cells for the development and establishment of cell therapy applications (e.g. genome editing generation of CAR-T cells)Generation of large numbers of transiently modified primary cells for cell-based assaysContent:22.5 mL P3 Primary Cell Nucleofector Solution5 mL Supplement 1,1 x LV Nucleocuvette Cartridge2 x 4D-Nucleofector LV Reservoir
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.