Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Terphenyl-d14 Solution, 2000μg/mL in methylene chloride, Ultra Scientific

Catalog No. USATS1601
Encompass
Click to view available options
Quantity:
1 x 1 mL

Chemical Identifiers

CAS 1718-51-0, 75-09-2
Molecular Formula C18H14
Molecular Weight (g/mol) 244.395
InChI Key XJKSTNDFUHDPQJ-WZAAGXFHSA-N
PubChem CID 123128
IUPAC Name 1,2,3,4,5-pentadeuterio-6-[2,3,5,6-tetradeuterio-4-(2,3,4,5,6-pentadeuteriophenyl)phenyl]benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=CC=C3

Specifications

Quantity 1 x 1 mL
CAS Max % 99.8, 0.15
Concentration or Composition (by Analyte or Components) 2000μg/mL
Analyte or Component Names p-terphenyl-d14,methylene chloride
Solvent or Matrix Methylene chloride
Chemical Name or Material p-Terphenyl-d14 Solution
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.