Learn More
New England Biolabs, Inc. p-Nitrophenylphosphate (PNPP) – 5 ml
Supplier: New England Biolabs, Inc. P0757L
p-Nitrophenyl Phosphate (PNPP) is a non-proteinaceous, non-specific substrate used to assay protein, alkaline and acid phosphatases.