Learn More
Eurofins Discoverx Oxytocin, 5 mg. (92-1060)
Supplier:  Eurofins Discoverx 921060
Encompass_Preferred
Oxytocin is a nine amino acid neurohypophysial hormone that acts as a neuromodulator in the brain. It is produced in the hypothalamus and secreted from the pituitary, and controls key parts of the reproductive system and brain, especially active during and after childbirth. Acts as an agonist for OXTR. Provided as 5 mg of dry powder. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9601
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.