Learn More
*TELEDYNE INSTRUMENTS OXYGEN SENSORS B1
Supplier:  *TELEDYNE INSTRUMENTS C06689B1
Encompass
General purpose percent oxygen analysis where fast response is desired (90% in less than 7 seconds).
Catalog No. NC0661313
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.