Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversey™ Oxivir™ 1 Wipes
Next generation ready-to-use, pre-wetted, one-step disinfectant cleaner wipe, based on proprietary AHP™ technology. Oxivir™ 1 Wipes are formulated to deliver broad spectrum disinfection in ONE minute or less, with 75 total claims.
Supplier: Diversey™ 100850925
This item is not returnable.
View return policy
Description
- 27.9 x 30.5 cm (11 x 12 in.) disinfectant wipes
- Broad spectrum, multi-surface cleaner/disinfectant and sanitizer
- Harnesses the cleaning power of hydrogen peroxide
- Specially designed to penetrate the toughest soils
- Fast, Effective, Responsible and Sustainable formulation
- Environmentally-responsible with no VOC's, no added fragrance, no NPE's
- Fast-acting, convenient formula, suitable for daily use, and contact time of ONE minute or less
- Formulated with patented AHP™ technology containing no phosphates, quats or phenols
- The active ingredient breaks down to water and oxygen after use
Specifications
Disinfectant Cleaner Wipes | |
White | |
160 Wipes/Pk., 4 Pk./Cs. | |
1.7 to 2.5 |
EPA, FDA | |
Bag | |
Non-sterile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction