Learn More
Saf T Pak Inc STP-500-OVPK Overpack System, Insulated, Category A, B and Exempt Patient Specimen, (UN 2814, UN 2900, UN 3373) Small, 10/Case
SDP
Supplier:  Saf T Pak Inc 35182
Encompass_Preferred
Overpack means an enclosure that is used by a single consignor to provide refrigeration, protection or convenience in the handling of a single fully compliant package or to consolidate two or more fully compliant packages. System Components:Outer Box OD 13.5 x 13.5 x 8.5", pre-printed with Overpack MarkingSTP-802 Class 6 Infectious Substance Label (1 ea.)STP-809 UN 2814 Infectious Substance Affecting Humans, Category A Label (1 ea.)STP-816 UN 2900 Infectious Substance Affecting Animals, Category A Label (1 ea.)STP-818 UN 3373 Marking Label (1 ea.)STP-819 Exempt Human/Animal Specimen Label (1 ea.)STP-820 Biological Substance, Category B Marking (1 ea.)
Catalog No. 50-190-4610
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.