Learn More
Volu Sol Acid Ethanol 90%, 128 oz / 3.78 L, Volu-Sol™
SDP
Supplier:  Volu Sol VXA128
Encompass_Preferred
Acid Ethanol 90, for use in qualitative procedures as a decolorizer in the Wheatley procedure for detection and identification of intestinal protozoa.  Component of Volu-Sol's Parasitology Kit
Catalog No. 50-420-867
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.