Learn More
World Precision Instrument Osteotomes - Bone Chisel, U.S. Army Pattern, 12mm
SDP
Supplier:  World Precision Instrument WP0319
Encompass_Preferred
This Osteotome, bone chisel, can be used for scraping, cleaning and sculpting bone. For example, you could chisel out bone fragments to access restricted spaces. Three tip sizes are available (6, 12, or 18mm), one for every application. This chisel is crafted from standard grade stainless steel, suitable for all forms of disinfection and autoclaving.
Catalog No. 50-253-8361
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.