Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Hepatitis B surface antigen (HBSAG)

Supplier: Ortho-Clinical Diagnostics™ 6801322
This item is not returnable.
View return policy
Specifications
Hepatitis B Antigen | |
Vitros™ HBsAg Reagent | |
100 Pack |
Well | |
Coated wells 100, conjugate reagent 6.2mL, assay reagent 8.4mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction