Learn More
Ortho-Clinical Diagnostics™ VITROS™ Carcinoembryonic antigen (CEA)
Supplier: Ortho-Clinical Diagnostics™ 1920115
This item is not returnable.
View return policy
Specifications
Carcinoembryonic Antigen | |
Vitros™ CEA Reagent | |
100 Pack | |
30 min. |
Well | |
Coated wells 100, Biotinylated Antibody reagent 9.7mL, conjugate reagent 9.7mL | |
20 μL |