Learn More
Ortho-Clinical Diagnostics™ VITROS™ Red Cell Folate Reagent
Supplier: Ortho-Clinical Diagnostics™ 1107804
This item is not returnable.
View return policy
Specifications
RBC Folate | |
Red Cell Folate Reagent | |
2 Bottles of Red Cell Folate reagent with 27mL antimicrobial agent, 2 Red Cell Folate Ascorbate Tablets (54mg Ascorbic Acid/tablet) individually foil-wrapped, protocol card for red cell folate (colored pale blue) |
EDTA Plasma, Heparin Plasma, Serum | |
Quantitative measurement of red cell folate | |
50 Tests |