Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ortho-Clinical Diagnostics™ VITROS™ Calibrators: Ferritin

Supplier: Ortho-Clinical Diagnostics™ 1158864
This item is not returnable.
View return policy
Specifications
2°C to 8°C | |
Human Serum and Plasma (Heparin) | |
VITROS™ Ferritin calibrators 1, 2 and 3 (freeze-dried, Human Spleen Ferritin in Human Plasma with antimicrobial agent, reconstitution volume 1mL), lot calibration card, protocol card, 24 calibrator bar code labels (8 for each calibrator) | |
3 | |
3 Sets/Pk. |
VITROS™ ECi/ECiQ immunodiagnostic system, 3600 immunodiagnostic system, 5600 integrated system | |
Ferritin Calibrator | |
Quantitative measurement of Ferritin | |
Yes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction