Learn More
McKesson General Medical Ora-Blend SF 16 oz. Citrus Berry Flavor
Supplier: McKesson General Medical 901859
Ora-Blend SF is a flavored sugar-free oral suspending vehicle used to simplify the process involved in the extemporaneous compounding of oral suspensions. The suspending agents form a structured, gel-like matrix which suspend particles and allow for little settling. Ora-Blend SF is buffered to a slightly acidic pH to help reduce degradation of medicinal agents through oxidation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.