Learn More
McKesson General Medical OPTICHAMBER DIAMOND 10/PK
Supplier: McKesson General Medical 1077478
Holding Chamber OptiChamber Diamond, 10/pack.Low resistance inspiratory and expiratory valves open freely, even under low pediatric pressures and flows.Soft-seal technology is designed to contour to face, reduce leakage with minimal applied force.Uniquely designed stepped mouthpiece facilitates transfer from pediatric mask to mouthpiece.Clear, anti-static chamber delivers consistent aerosol therapy and is ready to use right out of the package.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.