Learn More
Cole-Parmer Extech 461895 Combination Optical/contact Tachometer
Supplier: Cole-Parmer 8770007
These tachometers feature both contact and optical measurement capabilities. Memory button holds last reading up to 5 minutes and recalls minimum and maximum readings. Display flips with the unit to help identify measuring mode. Model 87700-10 features a laser sighting for spotting targets up to 6 1/2 feet (1.9 m) away.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.