Learn More
New England Biolabs, Inc. OneTaq Hot Start Quick-Load 2X Master Mix with Standard Buffer – 100 reactions (50 µl vol)
Supplier: New England Biolabs, Inc. M0488S
OneTaq Hot Start Quick-Load 2X Master Mix with Standard Buffer is an optimized, ready-to-use blend of Taq and Deep Vent DNA Polymerases combined with an aptamer-based inhibitor. This enzyme blend is ideally suited to routine PCR applications from a variety of templates, including pure DNA solutions, bacterial colonies and cDNA products. The 3' to 5' exonuclease activity of Deep Vent DNA Polymerase increases the fidelity and robust amplification of Taq DNA Polymerase. The hot start nature of the enzyme offers convenience with decreased interference from primer dimers and secondary products. The convenient Quick-Load master mix formulation contains dNTPs, MgCl2, buffer components and stabilizers as well as two commonly used tracking dyes for DNA gels. On a 1% agarose gel in 1X TBE, Xylene Cyanol FF migrates at ~4 kb and Tartrazine migrates at ~10 bp. Both dyes are present in concentrations that do not mask co-migrating DNA bands
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.